Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084405.2 | NP_001077874.1 | 4 - 5 | 935 | 122 | |
NM_001335188.1 | NP_001324121.1 | 4 - 5 | 1020 | 122 | ||
NM_001335189.1 | NP_001324119.1 | 4 - 5 | 1117 | 122 | ||
NM_001335190.1 | NP_001324120.1 | 3 - 4 | 1194 | 122 | ||
NM_126403.4 | NP_178451.2 | 5 - 6 | 943 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |