Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_102820.3 | NP_564364.1 | 6 - 7 | 894 | 100 | |
GenBank mRNA | AK118475.1 | - | 6 - 7 | 858 | 100 | |
AY084256.1 | - | 6 - 7 | 858 | 100 | ||
BT005308.1 | - | 6 - 7 | 696 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |