Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198198.1 | NP_001185127.1 | 14 - 15 | 1265 | 84 | |
NM_102984.5 | NP_174527.2 | 14 - 15 | 1290 | 84 | ||
GenBank mRNA | AY074318.1 | - | 14 - 15 | 1213 | 84 | |
AY133872.1 | - | 13 - 14 | 1067 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |