Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120040.4 | NP_195591.1 | 3 - 4 | 434 | 103 | |
GenBank mRNA | AF370297.1 | - | 3 - 4 | 361 | 103 | |
AY142681.1 | - | 3 - 4 | 254 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |