Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342522.1 | NP_001329800.1 | 1 - 2 | 498 | 97 | |
NM_120061.2 | NP_568050.1 | 1 - 2 | 581 | 97 | ||
GenBank mRNA | AY059825.1 | - | 1 - 2 | 498 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |