Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127039.4 | NP_179082.1 | 6 - 7 | 982 | 75 | |
GenBank mRNA | AY054287.1 | - | 6 - 7 | 829 | 75 | |
AY132010.1 | - | 6 - 7 | 737 | 75 | ||
U05238.1 | - | 6 - 7 | 769 | 75 | ||
X75782.1 | - | 778 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |