Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337082.1 | NP_001324514.1 | 7 - 8 | 589 | 65 | |
NM_001337083.1 | NP_001324515.1 | 7 - 8 | 497 | 65 | ||
NM_129998.3 | NP_566016.1 | 7 - 8 | 538 | 65 | ||
NM_180084.2 | NP_850415.1 | 7 - 8 | 541 | 65 | ||
GenBank mRNA | AF387018.1 | - | 7 - 8 | 540 | 65 | |
BT006613.1 | - | 6 - 7 | 406 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |