Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337133.1 | NP_001324015.1 | 2 - 3 | 1027 | 69 | |
NM_001337134.1 | NP_001324014.1 | 2 - 3 | 936 | 69 | ||
NM_180107.3 | NP_850438.2 | 3 - 4 | 929 | 69 | ||
GenBank mRNA | AB493593.1 | - | 2 - 3 | 699 | 69 | |
AY074647.1 | - | 2 - 3 | 703 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |