Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337538.1 | NP_001319470.1 | 3 - 4 | 818 | 86 | |
NM_111335.2 | NP_566233.1 | 4 - 5 | 639 | 86 | ||
NM_180183.3 | NP_850514.1 | 4 - 5 | 663 | 86 | ||
GenBank mRNA | AK117325.1 | - | 4 - 5 | 504 | 86 | |
BT024507.1 | - | 3 - 4 | 417 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |