Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337562.1 | NP_001325515.1 | 9 - 10 | 1245 | 75 | |
NM_111381.3 | NP_566246.1 | 9 - 10 | 1362 | 75 | ||
NM_180185.3 | NP_850516.1 | 9 - 10 | 1360 | 75 | ||
GenBank mRNA | AK226843.1 | - | 9 - 10 | 1320 | 75 | |
AY056292.1 | - | 9 - 10 | 1270 | 75 | ||
AY127022.1 | - | 9 - 10 | 1313 | 75 | ||
AY142674.1 | - | 8 - 9 | 961 | 75 | ||
BT000631.1 | - | 8 - 9 | 961 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |