Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112536.3 | NP_188285.1 | 7 - 8 | 1291 | 136 | |
NM_180267.2 | NP_850598.1 | 7 - 8 | 1269 | 136 | ||
GenBank mRNA | AK226569.1 | - | 7 - 8 | 1146 | 136 | |
AY056129.1 | - | 7 - 8 | 1127 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |