Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120202.5 | NP_195744.1 | 3 - 4 | 945 | 70 | |
NM_202990.2 | NP_974719.1 | 1 - 2 | 269 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |