Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120291.4 | NP_195833.1 | 5 - 6 | 699 | 77 | |
GenBank mRNA | BT025628.1 | - | 5 - 6 | 589 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |