Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342678.1 | NP_001331961.1 | 5 - 6 | 1309 | 142 | |
NM_120366.4 | NP_195908.1 | 5 - 6 | 1309 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |