Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202636.1 | NP_001189565.1 | 1 - 2 | 381 | 76 | |
NM_001202637.1 | NP_001189566.1 | 1 - 2 | 381 | 76 | ||
NM_127668.4 | NP_565495.1 | 1 - 2 | 463 | 76 | ||
GenBank mRNA | AB102951.1 | - | 1 - 2 | 106 | 76 | |
AJ617630.1 | - | 1 - 2 | 359 | 76 | ||
AY045836.1 | - | 1 - 2 | 366 | 76 | ||
AY087925.1 | - | 1 - 2 | 361 | 76 | ||
BT004371.1 | - | 1 - 2 | 106 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |