Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035776.1 | NP_001030853.1 | 12 - 13 | 1274 | 71 | |
NM_001339622.1 | NP_001327608.1 | 12 - 13 | 1222 | 71 | ||
NM_115217.3 | NP_190925.1 | 11 - 12 | 1293 | 71 | ||
NM_180364.4 | NP_850695.2 | 10 - 11 | 1502 | 71 | ||
NM_202696.3 | NP_974425.1 | 11 - 12 | 1247 | 71 | ||
GenBank mRNA | AK317353.1 | - | 12 - 13 | 1275 | 71 | |
D45354.1 | - | 11 - 12 | 1253 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |