Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 24 - 25 | 4012 | 68 | |
NM_180432.2 | NP_850763.1 | 23 - 24 | 3844 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |