Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120496.3 | NP_568134.1 | 9 - 10 | 1916 | 107 | |
NM_180432.2 | NP_850763.1 | 8 - 9 | 1748 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |