Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121446.5 | NP_196946.1 | 3 - 4 | 838 | 67 | |
NM_180487.4 | NP_850818.1 | 4 - 5 | 1026 | 67 | ||
NM_203050.3 | NP_974779.1 | 4 - 5 | 816 | 67 | ||
NM_203051.3 | NP_974780.1 | 4 - 5 | 865 | 67 | ||
GenBank mRNA | AY099597.1 | - | 3 - 4 | 797 | 67 | |
BT000249.1 | - | 3 - 4 | 662 | 67 | ||
DQ086862.1 | - | 3 - 4 | 662 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |