Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120636.4 | NP_196173.1 | 10 - 11 | 1193 | 65 | |
GenBank mRNA | AK118437.1 | - | 10 - 11 | 1169 | 65 | |
BT009685.1 | - | 10 - 11 | 1056 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |