Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342930.1 | NP_001331523.1 | 22 - 23 | 1672 | 82 | |
NM_120800.4 | NP_196335.2 | 24 - 25 | 2227 | 82 | ||
GenBank mRNA | AY244746.1 | - | 24 - 25 | 2057 | 82 | |
FJ708772.1 | - | 24 - 25 | 1872 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |