Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120865.5 | NP_196400.2 | 5 - 6 | 996 | 69 | |
GenBank mRNA | BT015345.1 | - | 5 - 6 | 777 | 69 | |
BT015800.1 | - | 4 - 5 | 597 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |