Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EME1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202643.1 | NP_001189572.1 | 8 - 9 | 1180 | 79 | |
NM_001335775.1 | NP_001323848.1 | 8 - 9 | 1236 | 79 | ||
NM_127749.5 | NP_179771.2 | 8 - 9 | 1248 | 79 | ||
GenBank mRNA | AK228007.1 | - | 8 - 9 | 1171 | 79 | |
BT006456.1 | - | 8 - 9 | 1130 | 79 | ||
FJ936556.1 | - | 8 - 9 | 1139 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |