Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCD2;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202647.1 | NP_001189576.1 | 2 - 3 | 677 | 91 | |
NM_127815.4 | NP_179835.1 | 2 - 3 | 699 | 91 | ||
GenBank mRNA | BT025242.1 | - | 2 - 3 | 405 | 91 | |
X83370.1 | - | 599 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |