Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT84B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127890.3 | NP_179907.1 | 1 | 1351 | 97 | |
GenBank mRNA | AK118431.1 | - | 1 | 1351 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |