Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036856.2 | NP_001031933.1 | 4 - 5 | 772 | 93 | |
NM_122277.2 | NP_197761.2 | 5 - 6 | 665 | 93 | ||
NM_180735.4 | NP_851066.2 | 5 - 6 | 754 | 93 | ||
GenBank mRNA | AB161693.1 | - | 5 - 6 | 721 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |