Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124787.4 | NP_200219.1 | 1 - 2 | 248 | 87 | |
NM_180856.4 | NP_851187.1 | 1 - 2 | 248 | 87 | ||
GenBank mRNA | AX417711.1 | - | 1 - 2 | 193 | 87 | |
AY140075.1 | - | 1 - 2 | 224 | 87 | ||
BT010329.1 | - | 1 - 2 | 193 | 87 |
Gene Symbol: AT5G51970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK230367.1 | - | 973 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |