Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125178.6 | NP_200601.3 | 2 - 3 | 243 | 115 | |
NM_180877.2 | NP_851208.1 | 2 - 3 | 222 | 115 | ||
GenBank mRNA | AX765838.1 | - | 2 - 3 | 145 | 115 | |
AY062701.1 | - | 2 - 3 | 222 | 115 | ||
AY114672.1 | - | 2 - 3 | 136 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |