Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125508.2 | NP_200923.2 | 9 - 10 | 1689 | 60 | |
NM_180906.2 | NP_851237.1 | 9 - 10 | 1689 | 60 | ||
GenBank mRNA | BT002058.1 | - | 9 - 10 | 1598 | 60 | |
BT008386.1 | - | 9 - 10 | 1492 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |