Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SC35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125816.4 | NP_201225.1 | 2 - 3 | 238 | 68 | |
NM_180930.3 | NP_851261.1 | 2 - 3 | 238 | 68 | ||
GenBank mRNA | AY063890.1 | - | 2 - 3 | 209 | 68 | |
AY122917.1 | - | 2 - 3 | 150 | 68 | ||
AY127004.1 | - | 2 - 3 | 227 | 68 | ||
AY143851.1 | - | 2 - 3 | 150 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |