Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203127.1 | NP_001190056.1 | 3 - 4 | 751 | 97 | |
NM_115022.3 | NP_566954.2 | 2 - 3 | 919 | 97 | ||
GenBank mRNA | AJ238802.1 | - | 2 - 3 | 912 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |