Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115342.5 | NP_191045.2 | 1 - 2 | 458 | 68 | |
GenBank mRNA | AY065279.1 | - | 1 - 2 | 380 | 68 | |
AY096530.1 | - | 1 - 2 | 287 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |