Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCG19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115371.3 | NP_191073.1 | 1 | 2124 | 72 | |
GenBank mRNA | AY045932.1 | - | 1 | 2062 | 72 | |
AY079387.1 | - | 1 | 1959 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |