Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSLA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115458.5 | NP_191159.2 | 7 - 8 | 1626 | 139 | |
GenBank mRNA | KJ138786.1 | - | 7 - 8 | 1425 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |