Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115462.4 | NP_567031.1 | 14 - 15 | 2497 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |