Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339858.1 | NP_001319779.1 | 12 - 13 | 1560 | 70 | |
NM_001339859.1 | NP_001325644.1 | 12 - 13 | 1560 | 70 | ||
NM_001339860.1 | NP_001325645.1 | 12 - 13 | 2325 | 70 | ||
NM_001339861.1 | NP_001325651.1 | 12 - 13 | 2379 | 70 | ||
NM_001339862.1 | NP_001325648.1 | 12 - 13 | 1994 | 70 | ||
NM_001339863.1 | NP_001325647.1 | 12 - 13 | 2032 | 70 | ||
NM_001339864.1 | NP_001325650.1 | 12 - 13 | 1540 | 70 | ||
NM_001339865.1 | NP_001325649.1 | 11 - 12 | 1696 | 70 | ||
NM_001339866.1 | NP_001325646.1 | 11 - 12 | 1734 | 70 | ||
NM_115601.3 | NP_567048.1 | 12 - 13 | 1598 | 70 | ||
GenBank mRNA | AK318626.1 | - | 11 - 12 | 1551 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |