Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124279.5 | NP_199713.2 | 11 - 12 | 1256 | 85 | |
NM_203184.2 | NP_974913.1 | 11 - 12 | 1190 | 85 | ||
GenBank mRNA | BT030319.1 | - | 11 - 12 | 1106 | 85 | |
BT030323.1 | - | 11 - 12 | 1106 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |