Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037031.1 | NP_001032108.1 | 8 - 9 | 823 | 76 | |
NM_001037032.2 | NP_001032109.2 | 8 - 9 | 823 | 76 | ||
NM_001125993.1 | NP_001119465.1 | 8 - 9 | 823 | 76 | ||
NM_001345415.1 | NP_001332003.1 | 8 - 9 | 853 | 76 | ||
NM_125434.4 | NP_200849.2 | 8 - 9 | 1009 | 76 | ||
NM_203240.3 | NP_974969.1 | 8 - 9 | 952 | 76 | ||
GenBank mRNA | AK175813.1 | - | 8 - 9 | 823 | 76 | |
AK220973.1 | - | 8 - 9 | 823 | 76 | ||
AK221190.1 | - | 8 - 9 | 823 | 76 | ||
AK221249.1 | - | 8 - 9 | 823 | 76 | ||
AK221292.1 | - | 8 - 9 | 823 | 76 | ||
AK316983.1 | - | 8 - 9 | 823 | 76 | ||
AY139752.1 | - | 8 - 9 | 822 | 76 | ||
AY700572.1 | - | 8 - 9 | 824 | 76 | ||
BT004542.1 | - | 7 - 8 | 585 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |