Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123934.1 | NP_001117406.1 | 4 - 5 | 659 | 104 | |
NM_103120.4 | NP_174660.2 | 4 - 5 | 680 | 104 | ||
GenBank mRNA | AY128383.1 | - | 4 - 5 | 640 | 104 | |
BT000112.1 | - | 4 - 5 | 527 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |