Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103495.4 | NP_564486.1 | 2 - 3 | 847 | 121 | |
GenBank mRNA | AY065453.1 | - | 2 - 3 | 800 | 121 | |
AY117284.1 | - | 2 - 3 | 726 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |