Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HXK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333285.1 | NP_001321728.1 | 8 - 9 | 1459 | 83 | |
NM_103682.4 | NP_175220.2 | 8 - 9 | 1314 | 83 | ||
GenBank mRNA | BT030472.1 | - | 8 - 9 | 1162 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |