Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333320.1 | NP_001321802.1 | 8 - 9 | 1065 | 66 | |
NM_103724.2 | NP_175261.1 | 8 - 9 | 1065 | 66 | ||
GenBank mRNA | AK228479.1 | - | 8 - 9 | 918 | 66 | |
U95143.1 | - | 8 - 9 | 1061 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |