Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339950.1 | NP_001326589.1 | 12 - 13 | 1567 | 87 | |
NM_115772.4 | NP_191469.3 | 18 - 19 | 2595 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |