Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARGOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115853.5 | NP_191549.2 | 1 | 682 | 81 | |
GenBank mRNA | AK176640.1 | - | 1 | 459 | 81 | |
BT011724.1 | - | 1 | 146 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |