Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121319.5 | NP_196820.1 | 3 - 4 | 868 | 100 | |
GenBank mRNA | AF314176.1 | - | 3 - 4 | 771 | 100 | |
AF372927.1 | - | 3 - 4 | 663 | 100 | ||
AY078037.1 | - | 3 - 4 | 520 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |