Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343274.1 | NP_001318555.1 | 8 - 9 | 1022 | 118 | |
NM_001343275.1 | NP_001331012.1 | 8 - 9 | 953 | 118 | ||
NM_001343276.1 | NP_001331011.1 | 8 - 9 | 953 | 118 | ||
NM_121351.5 | NP_196852.3 | 7 - 8 | 989 | 118 | ||
GenBank mRNA | BT012225.1 | - | 7 - 8 | 716 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |