Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121356.3 | NP_196857.2 | 7 - 8 | 2529 | 81 | |
GenBank mRNA | DQ315360.1 | - | 7 - 8 | 2113 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |