Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121413.2 | NP_196914.1 | 2 - 3 | 363 | 71 | |
GenBank mRNA | AK118738.1 | - | 2 - 3 | 252 | 71 | |
AY085446.1 | - | 2 - 3 | 363 | 71 | ||
BT006054.1 | - | 2 - 3 | 152 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |