Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336418.1 | NP_001325309.1 | 6 - 7 | 1242 | 111 | |
NM_128855.4 | NP_180854.1 | 6 - 7 | 1245 | 111 | ||
GenBank mRNA | BT025337.1 | - | 6 - 7 | 1126 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |