Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336434.1 | NP_001324212.1 | 28 - 29 | 4107 | 109 | |
NM_001336435.1 | NP_001324208.1 | 28 - 29 | 3674 | 109 | ||
NM_001336436.1 | NP_001324211.1 | 28 - 29 | 3674 | 109 | ||
NM_001336437.1 | NP_001324213.1 | 14 - 15 | 2146 | 109 | ||
NM_001336438.1 | NP_001324210.1 | 28 - 29 | 3935 | 109 | ||
NM_001336439.1 | NP_001324209.1 | 29 - 30 | 3611 | 109 | ||
NM_001336440.1 | NP_001324214.1 | 29 - 30 | 3611 | 109 | ||
NM_128883.3 | NP_180882.2 | 28 - 29 | 3674 | 109 | ||
GenBank mRNA | LC177513.1 | - | 27 - 28 | 3620 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |